Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | 2 | 611 | NP_001138872.1 |
NM_139174.3 | 3 | 827 | NP_631913.3 | |
XM_005255814.1 | 2 | 611 | XP_005255871.1 | |
XM_011522919.1 | 3 | 827 | XP_011521221.1 | |
XM_011522920.1 | 3 | 522 | XP_011521222.1 | |
AF447586.1 | 2 | 549 | AAM22869.1 | |
AK093046.1 | 3 | 806 | BAC04032.1 | |
AK093523.1 | 1 | 797 | ||
AK131102.1 | 3 | 3375 | BAC85152.1 | |
AK315171.1 | 2 | 530 | ||
BC033491.1 | 2 | 549 | AAH33491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | TAF1C DNAAF1 KCNG4 ADAD2 |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | TAF1C DNAAF1 HSDL1 MBTPS1 LOC729887 KCNG4 ADAD2 |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | TAF1C DNAAF1 LOC105371371 HSDL1 MBTPS1 NECAB2 LOC729887 SLC38A8 MLYCD KCNG4 ADAD2 OSGIN1 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | TAF1C DNAAF1 WFDC1 HSDL1 LOC105376777 MBTPS1 LOC729887 SLC38A8 KCNG4 ADAD2 |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | TAF1C DNAAF1 ADAD2 |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | TAF1C DNAAF1 WFDC1 LOC105376777 KCNG4 ADAD2 |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | TAF1C ADAD2 |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | TAF1C DNAAF1 KCNG4 ADAD2 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | DNAAF1 TLDC1 LOC101928474 MIR5093 LINC01081 CRISPLD2 LOC440390 KCNG4 ADAD2 TAF1C LOC101928557 WFDC1 FENDRR LOC105376777 FAM92B LOC105371374 FOXC2-AS1 LOC102724344 C16orf74 KIAA0513 MTHFSD COTL1 MIR1910 COX4I1 LOC146513 MIR6774 ATP2C2 LINC00311 IRF8 KLHL36 GINS2 LOC101928614 LINC01082 FOXC2 FOXF1 FLJ30679 USP10 LINC00917 EMC8 FOXL1 GSE1 LOC400548 ZDHHC7 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | TAF1C DNAAF1 HSDL1 MBTPS1 LOC729887 SLC38A8 KCNG4 ADAD2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |