Assay Details
Target Gene Details
Entrez Gene ID: | 4048 |
Gene Name: | leukotriene A4 hydrolase |
Gene Aliases: |
- |
Location: |
Chr.12:96000753-96043520 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LTA4H | NM_000895.2 | 19 | 1913 | NP_000886.1 |
NM_001256643.1 | 19 | 1844 | NP_001243572.1 | |
NM_001256644.1 | 18 | 1761 | NP_001243573.1 | |
NR_132659.1 | 19 | 1913 | ||
XM_005268871.1 | 18 | 1820 | XP_005268928.1 | |
AB209740.1 | 19 | 5888 | BAD92977.1 | |
AI582379.1 | 1 | 250 | ||
AK298017.1 | 18 | 1761 | ||
AK301308.1 | 19 | 1722 | ||
BC032528.1 | 19 | 1830 | AAH32528.1 | |
BC036555.1 | 6 | 1662 | AAH36555.1 | |
BX647158.1 | 19 | 1801 | ||
CR457068.1 | 19 | 1771 | CAG33349.1 | |
J02959.1 | 19 | 1801 | AAA36177.1 | |
J03459.1 | 19 | 1839 | AAA36176.1 | |
R41544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3630445 | Chr.12:95990852 - 96016216 on Build GRCh38 | Gain | HAL LTA4H |
nsv523292 | Chr.12:95992360 - 96015423 on Build GRCh38 | Gain | HAL LTA4H |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |