Assay Details
Target Gene Details
Entrez Gene ID: | 101927380 |
Gene Name: | C9orf41 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:74952965-74997382 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C9orf41-AS1 | NR_121183.1 | 5 | 950 |
Target Gene Details
Entrez Gene ID: | 138199 |
Gene Name: | carnosine N-methyltransferase 1 |
Gene Aliases: |
C9orf41, UPF0586 |
Location: |
Chr.9:74980794-75028456 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CARNMT1 | NM_001320497.1 | 6 | 819 | NP_001307426.1 |
NM_152420.2 | 6 | 1139 | NP_689633.1 | |
NR_135282.1 | 5 | 998 | ||
XM_005251701.3 | 6 | 814 | XP_005251758.2 | |
XM_017014269.1 | 6 | 1126 | XP_016869758.1 | |
XM_017014270.1 | 7 | 1072 | XP_016869759.1 | |
XM_017014271.1 | 7 | 1564 | XP_016869760.1 | |
AK098661.1 | 5 | 566 | BAC05369.1 | |
AK299691.1 | 5 | 841 | ||
BC034033.1 | 6 | 1103 | AAH34033.1 | |
BX538061.1 | 5 | 656 | CAD97992.1 | |
BX647386.1 | 6 | 908 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |