Assay Details
Target Gene Details
Entrez Gene ID: | 1068 |
Gene Name: | centrin 1 |
Gene Aliases: |
CEN1, CETN |
Location: |
Chr.18:580343-582020 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CETN1 | NM_004066.2 | 1 | 624 | NP_004057.1 |
BC029515.1 | 1 | 610 | AAH29515.1 | |
U03270.1 | 1 | 600 | AAC27343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv576236 | Chr.18:319176 - 794272 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576239 | Chr.18:394946 - 712037 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv5810n54 | Chr.18:351004 - 2238246 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576242 | Chr.18:567127 - 841320 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3641497 | Chr.18:551569 - 745897 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv528169 | Chr.18:551688 - 2240220 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3307n100 | Chr.18:469797 - 683607 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060188 | Chr.18:575087 - 863351 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv833577 | Chr.18:521331 - 725019 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3893055 | Chr.18:415105 - 743226 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)