Assay Details
Target Gene Details
Entrez Gene ID: | 51100 |
Gene Name: | SH3 domain containing GRB2 like endophilin B1 |
Gene Aliases: |
Bif-1, CGI-61, PPP1R70, dJ612B15.2 |
Location: |
Chr.1:86704570-86748184 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3GLB1 | NM_001206651.1 | 3 | 634 | NP_001193580.1 |
NM_001206652.1 | 3 | 634 | NP_001193581.1 | |
NM_001206653.1 | 2 | 492 | NP_001193582.1 | |
NM_016009.4 | 3 | 634 | NP_057093.1 | |
XM_006710672.2 | 3 | 634 | XP_006710735.1 | |
XM_006710673.2 | 2 | 462 | XP_006710736.1 | |
XM_011541538.1 | 3 | 634 | XP_011539840.1 | |
XM_017001408.1 | 2 | 462 | XP_016856897.1 | |
XM_017001409.1 | 3 | 634 | XP_016856898.1 | |
XM_017001410.1 | 3 | 634 | XP_016856899.1 | |
AB007960.2 | 3 | 599 | ||
AF151819.1 | 3 | 329 | AAD34056.1 | |
AF257318.1 | 3 | 329 | AAF81225.1 | |
AF263293.1 | 3 | 309 | AAF73017.1 | |
AF350371.1 | 3 | 385 | AAK27365.1 | |
AK001954.1 | 3 | 419 | BAA91999.1 | |
AK225864.1 | 3 | 454 | ||
AK303710.1 | 2 | 445 | ||
BC007455.2 | 3 | 349 | AAH07455.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |