Assay Details
Target Gene Details
Entrez Gene ID: | 1824 |
Gene Name: | desmocollin 2 |
Gene Aliases: |
ARVD11, CDHF2, DG2, DGII/III, DSC3 |
Location: |
Chr.18:31065974-31102432 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DSC2 | NM_004949.4 | 17 | 3383 | NP_004940.1 |
NM_024422.4 | 16 | 3337 | NP_077740.1 | |
XM_005258206.4 | 16 | 2838 | XP_005258263.1 | |
AK297029.1 | ||||
BC063291.1 | 16 | 3319 | AAH63291.1 | |
BE813598.1 | 1 | 134 | ||
BX648425.1 | 16 | 2889 | CAH56152.1 | |
CR749226.1 | 16 | 3056 | CAH18082.1 | |
X56807.1 | 17 | 2962 | CAA40141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2244 | Chr.18:31055588 - 31086414 on Build GRCh38 | Deletion | DSC2 |
More Information
Additional Information:
For this assay, SNP(s) [rs139923163,rs78190086] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic DGV Variation |