Assay Details
Target Gene Details
Entrez Gene ID: | 23587 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
C17orf81, DERP6, HSPC002, MST071, MSTP071 |
Location: |
Chr.17:7252053-7259940 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELP5 | NM_015362.4 | NP_056177.3 | ||
NM_203413.2 | 7 | 1153 | NP_981958.1 | |
NM_203414.2 | NP_981959.1 | |||
NM_203415.1 | NP_981960.1 | |||
XM_011523779.2 | XP_011522081.1 | |||
AB013910.1 | BAB87800.1 | |||
AF087868.1 | AAM10496.1 | |||
AF163262.1 | 6 | 872 | AAQ13591.1 | |
AK225983.1 | ||||
AK289940.1 | 6 | 1156 | ||
AL713741.1 | 2 | 497 | CAH56280.1 | |
BC002762.2 | AAH02762.2 | |||
BF921423.1 | ||||
BF982004.1 | ||||
BM970924.1 | ||||
CA431021.1 | ||||
CA450318.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv885e212 | Chr.17:7253579 - 7269312 on Build GRCh38 | Loss |
![]() ![]() |
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3639861 | Chr.17:7198042 - 7261345 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2665200 | Chr.17:7091816 - 7333062 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1963 | Chr.17:7244585 - 7333855 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)