Assay Details
Target Gene Details
Entrez Gene ID: | 604 |
Gene Name: | B-cell CLL/lymphoma 6 |
Gene Aliases: |
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51 |
Location: |
Chr.3:187721377-187745725 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCL6 | NM_001130845.1 | NP_001124317.1 | ||
NM_001134738.1 | NP_001128210.1 | |||
NM_001706.4 | NP_001697.2 | |||
XM_005247694.3 | XP_005247751.1 | |||
XM_011513062.2 | XP_011511364.1 | |||
AK304325.1 | ||||
AK308162.1 | ||||
BC142705.1 | ||||
BC150184.1 | ||||
BX649185.1 | CAE46203.1 | |||
EU139066.1 | ||||
EU883531.1 | ||||
S67779.1 | ||||
U00115.1 | AAC50054.1 | |||
Z21943.1 | CAA79937.1 |
Target Gene Details
Entrez Gene ID: | 100131635 |
Gene Name: | hCG1645011-like |
Gene Aliases: |
- |
Location: |
Chr.3:187702366-187732415 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100131635 | NR_034062.1 | |||
AW235368.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |