Assay Details
Target Gene Details
Entrez Gene ID: | 84167 |
Gene Name: | chromosome 19 open reading frame 44 |
Gene Aliases: |
- |
Location: |
Chr.19:16496311-16521369 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C19orf44 | NM_032207.3 | 9 | 2274 | NP_115583.1 |
XM_006722920.3 | 8 | 2178 | XP_006722983.1 | |
AK025395.1 | 9 | 2172 | BAB15125.1 | |
BC027869.1 | 9 | 2093 | AAH27869.1 |
Target Gene Details
Entrez Gene ID: | 10523 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
DAN16, SCAF6, SRA1 |
Location: |
Chr.19:16517889-16542452 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHERP | NM_006387.5 | 15 | 2488 | NP_006378.3 |
AB209730.1 | 6 | 865 | BAD92967.1 | |
AF536542.1 | 15 | 2455 | AAN77183.1 | |
AK296883.1 | 9 | 1199 | ||
AK298408.1 | 7 | 1216 | ||
BC021294.2 | 15 | 2473 | AAH21294.1 | |
U94836.2 | 14 | 2390 | AAB53327.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78607249] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |