Assay Details
Target Gene Details
Entrez Gene ID: | 3070 |
Gene Name: | helicase, lymphoid-specific |
Gene Aliases: |
ICF4, LSH, Nbla10143, PASG, SMARCA6 |
Location: |
Chr.10:94545767-94613905 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HELLS | NM_001289067.1 | 14 | 1700 | NP_001275996.1 |
NM_001289068.1 | 13 | 1517 | NP_001275997.1 | |
NM_001289069.1 | 13 | 1466 | NP_001275998.1 | |
NM_001289070.1 | 11 | 1268 | NP_001275999.1 | |
NM_001289071.1 | 14 | 1606 | NP_001276000.1 | |
NM_001289072.1 | 11 | 1172 | NP_001276001.1 | |
NM_001289073.1 | 12 | 1497 | NP_001276002.1 | |
NM_001289074.1 | 14 | 1481 | NP_001276003.1 | |
NM_001289075.1 | 12 | 1365 | NP_001276004.1 | |
NM_018063.4 | 13 | 1562 | NP_060533.2 | |
AB074174.1 | BAE45737.1 | |||
AB102716.1 | 14 | 1451 | BAD10844.1 | |
AB102717.1 | 12 | 1342 | BAD10845.1 | |
AB102718.1 | 13 | 1311 | BAD10846.1 | |
AB102719.1 | 11 | 1017 | BAD10847.1 | |
AB102720.1 | 12 | 1210 | BAD10848.1 | |
AB102721.1 | 11 | 1113 | BAD10849.1 | |
AB102722.1 | 14 | 1326 | BAD10850.1 | |
AB102723.1 | 14 | 1545 | BAD10851.1 | |
AB113248.1 | 13 | 1407 | BAD24804.1 | |
AB113249.1 | 13 | 1407 | BAD24805.1 | |
AF155827.1 | 13 | 1512 | AAF82262.1 | |
AK314485.1 | 13 | 1551 | ||
AY007108.1 | AAG01987.1 | |||
BC029381.1 | AAH29381.1 | |||
BC030963.1 | 2 | 165 | AAH30963.1 | |
BC031004.1 | 2 | 165 | AAH31004.1 | |
BX538033.1 | 13 | 1501 | CAD97978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517681 | Chr.10:94583471 - 94627322 on Build GRCh38 | Gain | HELLS |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |