Assay Details
Target Gene Details
Entrez Gene ID: | 8312 |
Gene Name: | axin 1 |
Gene Aliases: |
AXIN, PPP1R49 |
Location: |
Chr.16:287440-355226 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXIN1 | NM_003502.3 | NP_003493.1 | ||
NM_181050.2 | NP_851393.1 | |||
NR_134879.1 | ||||
XM_011522682.2 | XP_011520984.1 | |||
XM_011522683.2 | XP_011520985.1 | |||
XM_011522684.2 | XP_011520986.1 | |||
XM_011522686.1 | XP_011520988.1 | |||
XM_017023743.1 | XP_016879232.1 | |||
XM_017023744.1 | XP_016879233.1 | |||
XM_017023745.1 | XP_016879234.1 | |||
XM_017023746.1 | XP_016879235.1 | |||
XM_017023747.1 | XP_016879236.1 | |||
XM_017023748.1 | XP_016879237.1 | |||
AB208876.1 | BAD92113.1 | |||
AF009674.1 | AAC51624.1 | |||
BC044648.1 | AAH44648.1 | |||
KF615847.1 | ||||
KF615848.1 | ||||
KF615849.1 | ||||
KF615850.1 | ||||
KF615851.1 | ||||
KF615852.1 | ||||
KF615853.1 | ||||
KF615854.1 | ||||
KU178413.1 | ||||
KU178414.1 | ||||
KU178415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3637568 | Chr.16:300040 - 319624 on Build GRCh38 | Gain | AXIN1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR5587 MIR4516 C16orf59 ERVK13-1 UNKL LOC105371184 RAB11FIP3 DECR2 GFER PKD1 MSRB1 TPSAB1 IGFALS HBA2 LMF1-AS1 PRR35 MEIOB PDPK1 SPSB3 LOC729652 TELO2 RAB26 NOXO1 AXIN1 CASKIN1 C16orf91 PIGQ CAPN15 SNORA64 BAIAP3 GNPTG HS3ST6 TPSD1 DNASE1L2 DDX11L10 SYNGR3 E4F1 RNF151 KCTD5 NPRL3 ABCA17P MIR6511B1 TMEM8A TPSB2 PDIA2 PRR25 TBC1D24 HBZ TSC2 CCDC78 FAM234A RAB40C CACNA1H HBM LINC00254 HAGH WFIKKN1 RPS2 RNPS1 PRSS27 NDUFB10 HBA1 ABCA3 NME4 SNHG19 METTL26 SNORD60 SNHG9 UBE2I MSLN WASIR2 LUC7L TMEM204 MRPL28 SNORA10 TRAF7 NTHL1 POLR3K LOC105371046 MIR3177 WDR24 MIR3178 LOC105371038 SSTR5 MIR6859-4 FBXL16 RHBDF1 SOX8 HBQ1 LOC100134368 MIR6768 MIR1225 MCRIP2 LOC101929280 CEMP1 LOC100287175 ECI1 RHBDL1 RPUSD1 GNG13 LOC106660606 SSTR5-AS1 METRN IFT140 C1QTNF8 STUB1 LOC107984876 PTX4 MIR6767 MAPK8IP3 MPG NUBP2 ZNF598 FAM173A AMDHD2 SNORA78 MLST8 LOC107987233 JMJD8 HN1L LMF1 NTN3 RPL3L NPW MIR3677 RPL23AP5 LOC105371045 MIR4717 EME2 LOC105371049 MIR940 TBL3 FLJ42627 CRAMP1 MRPS34 CCNF ATP6V0C NME3 MIR662 HAGHL CLCN7 RGS11 LOC652276 CCDC154 CHTF18 LINC00235 BRICD5 LOC105371047 TSR3 RHOT2 TPSG1 ARHGDIG MIR3180-5 PGP PRSS29P SNRNP25 FAHD1 NARFL WDR90 SLC9A3R2 MIR3176 NHLRC4 |
dgv769e212 | Chr.16:301730 - 308308 on Build GRCh38 | Loss | AXIN1 |
esv3637564 | Chr.16:287941 - 370218 on Build GRCh38 | Gain | MRPL28 AXIN1 |
nsv952896 | Chr.16:273501 - 321500 on Build GRCh38 | Deletion | ARHGDIG PDIA2 RGS11 AXIN1 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | DECR2 LOC100134368 TMEM8A NME4 ARHGDIG PDIA2 MRPL28 RPL23AP5 RGS11 FAM234A RAB11FIP3 AXIN1 |
nsv570951 | Chr.16:279671 - 322597 on Build GRCh38 | Loss | ARHGDIG PDIA2 AXIN1 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | DECR2 LOC100134368 TMEM8A NME4 ARHGDIG PDIA2 MRPL28 RPL23AP5 RGS11 FAM234A RAB11FIP3 AXIN1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |