Assay Details
Target Gene Details
Entrez Gene ID: | 80018 |
Gene Name: | N(alpha)-acetyltransferase 25, NatB auxiliary subunit |
Gene Aliases: |
C12orf30, MDM20, NAP1 |
Location: |
Chr.12:112026689-112109022 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA25 | NM_024953.3 | 24 | 3071 | NP_079229.2 |
XM_005253938.1 | 23 | 2917 | XP_005253995.1 | |
XM_006719606.2 | 24 | 3731 | XP_006719669.1 | |
XM_017019977.1 | 22 | 2787 | XP_016875466.1 | |
AK023151.1 | BAB14432.1 | |||
AK292485.1 | ||||
AL832394.1 | 8 | 1139 | ||
BC113585.1 | ||||
BC113587.1 | ||||
BX641140.1 | 23 | 4167 | CAE46062.1 | |
BX647450.1 | 24 | 3024 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |