Assay Details
Target Gene Details
Entrez Gene ID: | 283232 |
Gene Name: | transmembrane protein 80 |
Gene Aliases: |
- |
Location: |
Chr.11:695386-705028 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM80 | NM_001042463.2 | NP_001035928.2 | ||
NM_001276253.1 | NP_001263182.1 | |||
NM_001276274.1 | NP_001263203.1 | |||
NM_174940.3 | NP_777600.3 | |||
XM_006718206.3 | XP_006718269.3 | |||
XM_017017600.1 | XP_016873089.1 | |||
AK124309.1 | 2 | 1343 | ||
AK302585.1 | ||||
BC008671.1 | ||||
BI916171.1 | ||||
BM874316.1 | ||||
CF134831.1 | ||||
CK819789.1 | ||||
DR006597.1 | ||||
HY080028.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625074 | Chr.11:669311 - 702135 on Build GRCh38 | Loss | TMEM80 DEAF1 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | ANO9 RASSF7 MUC5B PANO1 PHRF1 CRACR2B DRD4 PTDSS2 LOC101927503 SLC25A22 SNORA52 RNH1 PKP3 MIR210HG MIR210 CDHR5 CD151 LOC101059906 PIDD1 TSPAN4 PDDC1 RPLP2 LOC143666 B4GALNT4 HRAS SIGIRR TOLLIP TALDO1 CHID1 EPS8L2 POLR2L MUC2 SCT LRRC56 LMNTD2 MUC5AC MIR6744 IRF7 PNPLA2 TMEM80 MUC6 CEND1 LOC171391 AP2A2 DEAF1 |
nsv832043 | Chr.11:631027 - 758602 on Build GRCh38 | Loss | DRD4 TMEM80 TALDO1 EPS8L2 DEAF1 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | ANO9 RASSF7 MUC5B PANO1 PHRF1 CRACR2B DRD4 PTDSS2 LOC101927503 SLC25A22 SNORA52 RNH1 MIR210HG MIR210 CDHR5 CD151 LOC101059906 PIDD1 TSPAN4 PDDC1 RPLP2 LOC143666 HRAS SIGIRR TOLLIP TALDO1 CHID1 EPS8L2 POLR2L MUC2 SCT LRRC56 LMNTD2 MUC5AC MIR6744 IRF7 PNPLA2 TMEM80 MUC6 TOLLIP-AS1 CEND1 LOC171391 AP2A2 DEAF1 |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion | ANO9 RASSF7 PANO1 PHRF1 CRACR2B DRD4 PTDSS2 SLC25A22 SNORA52 RNH1 PKP3 MIR210HG MIR210 CDHR5 CD151 LOC101059906 PIDD1 TSPAN4 PDDC1 RPLP2 LOC143666 B4GALNT4 HRAS SIGIRR TALDO1 CHID1 EPS8L2 POLR2L SCT LRRC56 LMNTD2 IRF7 PNPLA2 TMEM80 CEND1 LOC171391 AP2A2 DEAF1 |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion | RASSF7 PANO1 PHRF1 CRACR2B DRD4 PTDSS2 SLC25A22 SNORA52 RNH1 MIR210HG MIR210 CDHR5 CD151 LOC101059906 PIDD1 TSPAN4 PDDC1 RPLP2 LOC143666 HRAS TALDO1 CHID1 EPS8L2 POLR2L SCT LRRC56 LMNTD2 IRF7 PNPLA2 TMEM80 CEND1 LOC171391 DEAF1 |