Assay Details
Target Gene Details
Entrez Gene ID: | 101926934 |
Gene Name: | uncharacterized LOC101926934 |
Gene Aliases: |
- |
Location: |
Chr.6:46670444-46688165 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101926934 | NR_134642.1 | |||
NR_134643.1 | ||||
NR_134644.1 | ||||
AA725776.1 | ||||
AI638322.1 | ||||
AI698692.1 | ||||
BX114358.1 |
Target Gene Details
Entrez Gene ID: | 9481 |
Gene Name: | solute carrier family 25 member 27 |
Gene Aliases: |
UCP4 |
Location: |
Chr.6:46652915-46678190 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A27 | NM_001204051.1 | NP_001190980.1 | ||
NM_001204052.1 | NP_001190981.1 | |||
NM_004277.4 | NP_004268.3 | |||
XM_005249483.2 | XP_005249540.1 | |||
XM_017011512.1 | XP_016867001.1 | |||
AF110532.1 | AAD16995.1 | |||
AK090871.1 | ||||
AK291427.1 | ||||
AK295233.1 | ||||
AK302906.1 | ||||
AK315940.1 | ||||
AY358711.1 | AAQ89951.1 | |||
BC033091.1 | AAH33091.1 | |||
CB243903.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |