Assay Details
Target Gene Details
Entrez Gene ID: | 4750 |
Gene Name: | NIMA related kinase 1 |
Gene Aliases: |
NY-REN-55, SRPS2, SRPS2A, SRTD6 |
Location: |
Chr.4:169393270-169612629 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEK1 | NM_001199397.1 | 33 | 4119 | NP_001186326.1 |
NM_001199398.1 | 32 | 3987 | NP_001186327.1 | |
NM_001199399.1 | 30 | 3828 | NP_001186328.1 | |
NM_001199400.1 | 30 | 3981 | NP_001186329.1 | |
NM_012224.2 | 31 | 4113 | NP_036356.1 | |
XM_006714228.1 | 32 | 3944 | XP_006714291.1 | |
XM_011532003.1 | 32 | 4037 | XP_011530305.1 | |
XM_011532004.1 | 31 | 3905 | XP_011530306.1 | |
XM_017008249.1 | 32 | 4023 | XP_016863738.1 | |
XM_017008250.1 | 31 | 4141 | XP_016863739.1 | |
XM_017008251.1 | 31 | 3939 | XP_016863740.1 | |
XM_017008252.1 | 29 | 3959 | XP_016863741.1 | |
XM_017008253.1 | 32 | 3969 | XP_016863742.1 | |
XM_017008254.1 | 25 | 3079 | XP_016863743.1 | |
AB067488.1 | 31 | 3949 | ||
AF155113.1 | 15 | 1997 | AAD42879.1 | |
AK025658.1 | 6 | 867 | BAB15207.1 | |
AL050385.1 | 32 | 3943 | CAI46225.1 | |
BC037790.2 | 5 | 779 | AAH37790.1 | |
BC114491.1 | 30 | 3588 | ||
CD358162.1 | 1 | 118 | ||
CR933642.1 | 33 | 4077 | CAI45943.1 | |
CR936672.1 | 30 | 3939 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |