Assay Details
Target Gene Details
Entrez Gene ID: | 1742 |
Gene Name: | discs large MAGUK scaffold protein 4 |
Gene Aliases: |
PSD95, SAP-90, SAP90 |
Location: |
Chr.17:7189890-7220050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLG4 | NM_001128827.2 | NP_001122299.1 | ||
NM_001321074.1 | NP_001308003.1 | |||
NM_001321075.1 | NP_001308004.1 | |||
NM_001321076.1 | NP_001308005.1 | |||
NM_001321077.1 | NP_001308006.1 | |||
NM_001365.4 | NP_001356.1 | |||
NR_135527.1 | ||||
XM_005256491.1 | XP_005256548.1 | |||
XM_011523698.1 | XP_011522000.1 | |||
XM_011523699.2 | XP_011522001.1 | |||
XM_011523702.1 | XP_011522004.1 | |||
XM_017024288.1 | XP_016879777.1 | |||
XM_017024289.1 | XP_016879778.1 | |||
XM_017024290.1 | XP_016879779.1 | |||
AK293835.1 | ||||
AK296357.1 | ||||
AK297346.1 | ||||
AK299801.1 | ||||
AK308533.1 | ||||
AK316096.1 | ||||
BC040533.1 | AAH40533.1 | |||
BC136553.1 | ||||
U83192.1 | AAC52113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | ALOX12 SLC16A11 TNK1 SHBG TEKT1 SPEM1 GPS2 MIR324 NLGN2 ZBTB4 ATP1B2 RNASEK-C17orf49 SLC16A13 PLSCR3 ASGR2 SENP3 CLEC10A YBX2 TNFSF12 EIF5A SOX15 SAT2 ASGR1 CHRNB1 LOC107983988 SNORA67 GABARAP RNASEK MIR195 BCL6B DLG4 ALOX12-AS1 SNORD10 MIR497 ACADVL LOC100996842 TMEM256 C17orf49 FGF11 PHF23 SENP3-EIF4A1 MPDU1 ALOX12P2 DVL2 TMEM256-PLSCR3 NEURL4 FXR2 CD68 C17orf74 ACAP1 TNFSF13 CLDN7 MIR497HG TNFSF12-TNFSF13 EIF4A1 FBXO39 TMEM95 KCTD11 ELP5 SNORA48 TMEM102 POLR2A SLC35G6 SLC2A4 CTDNEP1 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | TMEM256 TNK1 SPEM1 FGF11 PHF23 GPS2 MIR324 NLGN2 DVL2 TMEM256-PLSCR3 NEURL4 PLSCR3 YBX2 C17orf74 ACAP1 EIF5A CLDN7 ASGR1 CHRNB1 LOC107983988 TMEM95 GABARAP KCTD11 ELP5 DLG4 TMEM102 SLC2A4 CTDNEP1 ACADVL |
esv2665200 | Chr.17:7091816 - 7333062 on Build GRCh38 | Deletion | ASGR1 PHF23 GPS2 GABARAP MIR324 ELP5 DVL2 DLG4 NEURL4 ASGR2 YBX2 SLC2A4 CTDNEP1 EIF5A ACADVL CLDN7 |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | ALOX12 SLC16A11 TNK1 SHBG SPEM1 GPS2 MIR324 NLGN2 ZBTB4 ATP1B2 RNASEK-C17orf49 SLC16A13 PLSCR3 ASGR2 SENP3 CLEC10A YBX2 TNFSF12 WRAP53 EIF5A SOX15 SAT2 ASGR1 CHRNB1 LOC107983988 SNORA67 GABARAP RNASEK MIR195 BCL6B DLG4 ALOX12-AS1 SNORD10 MIR497 ACADVL LOC100996842 TMEM256 C17orf49 FGF11 PHF23 SENP3-EIF4A1 MPDU1 TP53 DVL2 TMEM256-PLSCR3 NEURL4 FXR2 CD68 C17orf74 ACAP1 TNFSF13 CLDN7 MIR497HG TNFSF12-TNFSF13 EIF4A1 TMEM95 KCTD11 ELP5 SNORA48 TMEM102 POLR2A SLC35G6 SLC2A4 CTDNEP1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |