Assay Details
Target Gene Details
Entrez Gene ID: | 10693 |
Gene Name: | chaperonin containing TCP1 subunit 6B |
Gene Aliases: |
CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303 |
Location: |
Chr.17:34927859-34981221 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCT6B | NM_001193529.2 | 13 | 1622 | NP_001180458.1 |
NM_001193530.1 | 13 | 1598 | NP_001180459.1 | |
NM_006584.3 | 14 | 1733 | NP_006575.2 | |
XM_011524203.2 | 12 | 1352 | XP_011522505.1 | |
XM_017024023.1 | 14 | 1636 | XP_016879512.1 | |
XM_017024024.1 | 11 | 1493 | XP_016879513.1 | |
AK225728.1 | 14 | 1654 | ||
AK225729.1 | 13 | 1515 | ||
AK301773.1 | 13 | 1511 | ||
AK302344.1 | 13 | 1545 | ||
BC026125.2 | 14 | 1626 | AAH26125.1 | |
BC027591.1 | 14 | 1713 | AAH27591.1 | |
BI825786.1 | 4 | 315 | ||
CR933688.1 | 8 | 3262 | CAI45976.1 | |
D78333.1 | 14 | 1658 | BAA11347.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |