Assay Details
Target Gene Details
Entrez Gene ID: | 9117 |
Gene Name: | SEC22 homolog C, vesicle trafficking protein |
Gene Aliases: |
SEC22L3 |
Location: |
Chr.3:42547967-42601080 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEC22C | NM_001201572.1 | 7 | 1345 | NP_001188501.1 |
NM_001201584.1 | 6 | 1341 | NP_001188513.1 | |
NM_004206.3 | 7 | 1407 | NP_004197.1 | |
NM_032970.3 | 7 | 6198 | NP_116752.1 | |
XM_005265533.1 | 8 | 6372 | XP_005265590.1 | |
XM_005265534.1 | 8 | 6272 | XP_005265591.1 | |
XM_005265535.1 | 8 | 6330 | XP_005265592.1 | |
XM_011534212.1 | 7 | 6050 | XP_011532514.1 | |
XM_017007438.1 | 8 | 1581 | XP_016862927.1 | |
XM_017007439.1 | 8 | 1601 | XP_016862928.1 | |
AK074576.1 | 7 | 1406 | ||
AK293133.1 | ||||
AY359041.1 | 6 | 1267 | AAQ89400.1 | |
BC018437.1 | 7 | 1315 | AAH18437.1 | |
CB410223.1 | 1 | 242 | ||
CR749670.1 | 5 | 5752 | CAH18461.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |