Assay Details
Target Gene Details
Entrez Gene ID: | 80832 |
Gene Name: | apolipoprotein L4 |
Gene Aliases: |
APOL-IV, APOLIV |
Location: |
Chr.22:36189128-36204833 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOL4 | NM_030643.4 | 6 | 726 | NP_085146.2 |
NM_145660.2 | 5 | 537 | NP_663693.1 | |
AF305226.1 | 4 | 378 | AAK20212.1 | |
AK225696.1 | 4 | 387 | ||
AK290466.1 | 5 | 711 | ||
AK290777.1 | 4 | 358 | ||
AK311506.1 | 5 | 510 | ||
AK312716.1 | 4 | 436 | ||
AK313637.1 | 5 | 474 | ||
AY014914.1 | 6 | 727 | AAG50352.1 | |
AY014915.1 | 5 | 537 | AAG50353.1 | |
CR456380.1 | 4 | 316 | CAG30266.1 | |
DA865632.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78582347] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |