Assay Details
Target Gene Details
Entrez Gene ID: | 9797 |
Gene Name: | TatD DNase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:10248493-10281222 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TATDN2 | NM_014760.3 | 4 | 1820 | NP_055575.3 |
AK289794.1 | 4 | 1541 | ||
BC090935.1 | 4 | 1401 | AAH90935.1 | |
BC101770.1 | 4 | 1309 | AAI01771.1 | |
BC101776.1 | 4 | 1310 | AAI01777.1 | |
D86972.1 | 5 | 1637 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | TAMM41 SLC6A1-AS1 BRK1 ATP2B2-IT2 LINC00852 SLC6A1 LINC00606 SEC13 IRAK2 SLC6A11 HRH1 MIR378B VGLL4 GHRLOS GHRL FANCD2OS LOC101927467 MIR885 VHL ATG7 FANCD2 TATDN2 ATP2B2 |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | GHRLOS GHRL LINC00852 TATDN2 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | TAMM41 SLC6A1-AS1 CIDECP ATP2B2-IT2 LINC00852 SLC6A1 LINC00606 LOC401052 EMC3 SLC6A11 HRH1 VGLL4 GHRLOS GHRL LOC101927467 VHL ATG7 BRK1 SEC13 FANCD2P2 IRAK2 MIR378B FANCD2OS MIR885 FANCD2 TATDN2 ATP2B2 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | TAMM41 SLC6A1-AS1 BRK1 ATP2B2-IT2 LINC00852 SLC6A1 LINC00606 SEC13 FANCD2P2 IRAK2 SLC6A11 HRH1 MIR378B VGLL4 GHRLOS GHRL FANCD2OS LOC101927467 MIR885 VHL ATG7 FANCD2 TATDN2 ATP2B2 |
nsv236904 | Chr.3:10266949 - 10274997 on Build GRCh38 | Deletion | TATDN2 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | TAMM41 SLC6A1-AS1 BRK1 ATP2B2-IT2 LINC00852 SLC6A1 LINC00606 SEC13 FANCD2P2 IRAK2 SLC6A11 HRH1 MIR378B VGLL4 GHRLOS GHRL FANCD2OS LOC101927467 MIR885 VHL ATG7 FANCD2 TATDN2 ATP2B2 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | TAMM41 SLC6A1-AS1 CIDECP ATP2B2-IT2 LINC00852 SLC6A1 LINC00606 LOC401052 EMC3 SLC6A11 HRH1 VGLL4 GHRLOS GHRL LOC101927467 VHL ATG7 EMC3-AS1 BRK1 SEC13 FANCD2P2 IRAK2 MIR378B FANCD2OS MIR885 FANCD2 TATDN2 ATP2B2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |