Assay Details
Target Gene Details
Entrez Gene ID: | 89876 |
Gene Name: | MYCBP associated and testis expressed 1 |
Gene Aliases: |
AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26 |
Location: |
Chr.3:119703022-119767102 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAATS1 | NM_001320316.1 | NP_001307245.1 | ||
NM_001320317.1 | NP_001307246.1 | |||
NM_001320318.1 | NP_001307247.1 | |||
NM_033364.3 | NP_203528.2 | |||
XM_011513277.1 | XP_011511579.1 | |||
AF440403.1 | AAP97317.1 | |||
AF497717.1 | AAM19218.1 | |||
AK055558.1 | ||||
AK289914.1 | ||||
AK294413.1 | ||||
AK302699.1 | ||||
AK303997.1 | ||||
AK309317.1 | ||||
AL133066.1 | CAB61389.1 | |||
BC035248.1 | AAH35248.1 | |||
BC126394.1 | ||||
CR749242.1 | CAH18098.1 | |||
DB445414.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv591391 | Chr.3:119684298 - 119723725 on Build GRCh38 | Loss | MAATS1 |
nsv591390 | Chr.3:119665316 - 119740004 on Build GRCh38 | Gain | COX17 MAATS1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |