Assay Details
Target Gene Details
Entrez Gene ID: | 64285 |
Gene Name: | rhomboid 5 homolog 1 |
Gene Aliases: |
C16orf8, Dist1, EGFR-RS, gene-89, gene-90, hDist1 |
Location: |
Chr.16:58059-76355 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RHBDF1 | NM_022450.3 | NP_071895.3 | ||
XM_005255494.1 | XP_005255551.1 | |||
XM_005255498.2 | XP_005255555.1 | |||
XM_006720921.1 | XP_006720984.1 | |||
XM_017023556.1 | XP_016879045.1 | |||
XM_017023557.1 | XP_016879046.1 | |||
XM_017023558.1 | 12 | 2038 | XP_016879047.1 | |
AK026010.1 | BAB15318.1 | |||
AK056708.1 | 12 | 2721 | ||
AK225408.1 | ||||
AK291177.1 | ||||
BC014425.1 | AAH14425.1 | |||
M99624.1 | AAA02490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518291 | Chr.16:47354 - 254515 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1044732 | Chr.16:35815 - 60707 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv2690n100 | Chr.16:10765 - 207261 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv988339 | Chr.16:56481 - 68287 on Build GRCh38 | Deletion |
![]() ![]() |
esv2760065 | Chr.16:31052 - 143601 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)