Assay Details
Target Gene Details
Entrez Gene ID: | 26268 |
Gene Name: | F-box protein 9 |
Gene Aliases: |
FBX9, NY-REN-57, VCIA1, dJ341E18.2 |
Location: |
Chr.6:53057807-53100873 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXO9 | NM_012347.4 | NP_036479.1 | ||
NM_033480.2 | NP_258441.1 | |||
NM_033481.3 | NP_258442.2 | |||
XM_005248995.4 | XP_005249052.1 | |||
XM_005248996.4 | XP_005249053.1 | |||
AB209833.1 | BAD93070.1 | |||
AF174597.1 | AAF04518.1 | |||
AF176704.1 | AAF03704.1 | |||
AK021559.1 | ||||
AK095307.1 | 2 | 1783 | ||
AK095315.1 | 1 | 2906 | ||
AL137520.1 | CAB70786.2 | |||
AY858798.1 | AAW51115.1 | |||
BC000650.2 | AAH00650.2 | |||
BX641082.1 | 2 | 738 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |