Assay Details
Target Gene Details
Entrez Gene ID: | 53635 |
Gene Name: | prostate tumor overexpressed 1 |
Gene Aliases: |
ACID2, PTOV-1 |
Location: |
Chr.19:49850726-49860744 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTOV1 | NM_001305105.1 | NP_001292034.1 | ||
NM_001305108.1 | NP_001292037.1 | |||
NM_017432.4 | NP_059128.2 | |||
NR_130963.1 | ||||
XM_005258998.4 | XP_005259055.1 | |||
XM_006723244.3 | XP_006723307.1 | |||
XM_011527033.1 | XP_011525335.1 | |||
XM_011527034.2 | XP_011525336.1 | |||
XM_011527035.1 | XP_011525337.1 | |||
XM_017026879.1 | XP_016882368.1 | |||
XM_017026880.1 | XP_016882369.1 | |||
AA582677.1 | ||||
AF218026.1 | AAG17268.1 | |||
AJ617480.1 | CAE84582.1 | |||
AK057817.1 | ||||
AY358168.1 | AAQ88535.1 | |||
BC015172.1 | AAH15172.1 | |||
U79287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | PTOV1-AS1 TULP2 MIR6798 MIR4324 SNAR-C5 SIGLEC11 SNAR-A7 ATF5 LIG1 SNAR-B1 SNAR-D CABP5 SNAR-G2 SPACA4 BCAT2 SNORD35B FTL SNAR-A6 GRWD1 ALDH16A1 LMTK3 RPL18 SNAR-A5 SNAR-A14 MIR4750 LIN7B ADM5 SYNGR4 CGB7 PLEKHA4 KCNA7 DKKL1 CARD8-AS1 IL4I1 IZUMO1 SEC1P GYS1 MIR6800 BCL2L12 MED25 NUP62 C19orf68 CCDC114 SNAR-A13 CGB8 MIR150 IRF3 SNAR-A4 ZNF473 RRAS SIGLEC16 PLA2G4C MIR6799 RUVBL2 TMEM143 KCNJ14 PIH1D1 SNAR-A11 LOC105447645 MAMSTR SNORD34 SNAR-A9 RPS11 PRMT1 SNAR-A12 ZNF114 VRK3 SNORD33 PPP1R15A MIR4749 NTF4 LOC105372430 NUCB1-AS1 ELSPBP1 CPT1C C19orf73 GFY CGB5 FUT1 MIR5088 NTN5 SNAR-C3 BSPH1 TEAD2 LOC100287477 BAX RPL13A PNKP CA11 SLC6A16 HRC SPHK2 SNAR-C2 TSKS CARD8 LOC101928295 PRR12 RASIP1 FLJ26850 GRIN2D SNAR-A8 SNAR-A3 FGF21 SCAF1 CD37 PTH2 CGB3 CGB1 PTOV1 SNAR-C4 SNRNP70 FUZ NUCB1 SNAR-C1 DBP PTOV1-AS2 FLT3LG EMP3 PLA2G4C-AS1 LHB SNAR-A2 MIR4751 AKT1S1 RCN3 FAM83E SNAR-A1 AP2A1 HSD17B14 CCDC155 CYTH2 SNAR-G1 SLC17A7 NOSIP TBC1D17 TRPM4 LOC101059948 SNAR-A10 SULT2B1 DHDH PPFIA3 SNORD32A FUT2 SNORD35A PRRG2 CGB2 SNAR-B2 FCGRT KDELR1 |
nsv526372 | Chr.19:49853907 - 49877601 on Build GRCh38 | Loss | PTOV1-AS2 PTOV1 MIR4749 PNKP AKT1S1 TBC1D17 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | AP2A1 PTOV1 MIR4749 PTOV1-AS1 PNKP MED25 NUP62 ATF5 FUZ MIR4750 TBC1D17 TSKS PTOV1-AS2 MIR6799 IL4I1 MIR4751 AKT1S1 MIR6800 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | PTOV1 MIR4749 PTOV1-AS1 BCL2L12 MED25 NUP62 ATF5 FUZ CPT1C PTOV1-AS2 IRF3 MIR5088 RRAS MIR6799 MIR4751 AKT1S1 AP2A1 PNKP NOSIP MIR4750 TBC1D17 ADM5 TSKS PRR12 PRMT1 IL4I1 PRRG2 SCAF1 MIR6800 |
dgv54n68 | Chr.19:49848512 - 50008788 on Build GRCh38 | Loss | PTOV1 MIR4749 PTOV1-AS1 PNKP NUP62 SIGLEC11 ATF5 MIR4750 TBC1D17 PTOV1-AS2 SIGLEC16 IL4I1 VRK3 MIR4751 AKT1S1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |