Assay Details
Target Gene Details
Entrez Gene ID: | 55010 |
Gene Name: | PARP1 binding protein |
Gene Aliases: |
AROM, C12orf48, PARI |
Location: |
Chr.12:102120171-102197520 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARPBP | NM_001319988.1 | 12 | 2955 | NP_001306917.1 |
NM_001319993.1 | 12 | 2832 | NP_001306922.1 | |
NM_001319994.1 | 11 | 2719 | NP_001306923.1 | |
NM_017915.4 | 11 | 2724 | NP_060385.3 | |
NR_135123.1 | 5 | 1804 | ||
XM_011538509.1 | 10 | 2490 | XP_011536811.1 | |
XM_011538514.2 | 10 | 2537 | XP_011536816.1 | |
XM_011538515.1 | 9 | 2330 | XP_011536817.1 | |
XM_017019534.1 | 10 | 2553 | XP_016875023.1 | |
XM_017019535.1 | 10 | 2563 | XP_016875024.1 | |
XM_017019536.1 | 9 | 2466 | XP_016875025.1 | |
XM_017019537.1 | 9 | 2361 | XP_016875026.1 | |
XM_017019538.1 | 10 | 2588 | XP_016875027.1 | |
XM_017019539.1 | 8 | 2127 | XP_016875028.1 | |
XM_017019540.1 | 9 | 2323 | XP_016875029.1 | |
XM_017019542.1 | 8 | 2330 | XP_016875031.1 | |
XM_017019543.1 | 8 | 2219 | XP_016875032.1 | |
XM_017019544.1 | 7 | 2140 | XP_016875033.1 | |
AA757264.1 | 1 | 368 | ||
AK000648.1 | BAA91306.1 | |||
BC098313.1 | 8 | 2114 | AAH98313.1 | |
BC099734.1 | 6 | 1928 | ||
BC099844.1 | 7 | 2007 | ||
CA438709.1 | 1 | 382 |
Target Gene Details
Entrez Gene ID: | 5367 |
Gene Name: | pro-melanin concentrating hormone |
Gene Aliases: |
MCH, ppMCH |
Location: |
Chr.12:102196459-102199540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PMCH | NM_002674.3 | NP_002665.2 | ||
XM_017019483.1 | XP_016874972.1 | |||
BC018048.1 | AAH18048.1 | |||
CB047052.1 | ||||
CB047053.1 | ||||
M57703.1 | AAA63214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv455702 | Chr.12:102163421 - 102233833 on Build GRCh38 | Loss | PARPBP PMCH |
More Information
Set Membership: |
Intragenic Exonic Non-exonic DGV Variation |