Assay Details
Target Gene Details
Entrez Gene ID: | 283337 |
Gene Name: | zinc finger protein 740 |
Gene Aliases: |
Zfp740 |
Location: |
Chr.12:53180715-53190870 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF740 | NM_001004304.3 | NP_001004304.1 | ||
XM_006719344.2 | XP_006719407.1 | |||
XM_011538194.2 | XP_011536496.1 | |||
XM_017019181.1 | XP_016874670.1 | |||
AK091946.1 | ||||
AK292744.1 | ||||
BC053557.1 | AAH53557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | CSAD IGFBP6 MFSD5 RARG SOAT2 SPRYD3 ZNF740 ITGB7 |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | CSAD ESPL1 MFSD5 RARG SOAT2 ZNF740 ITGB7 |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | CSAD IGFBP6 RARG SOAT2 ZNF740 ITGB7 |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | SP7 ESPL1 AAAS IGFBP6 TNS2 SOAT2 EIF4B CSAD LOC283335 PFDN5 MFSD5 RARG MIR6757 C12orf10 SPRYD3 ZNF740 ITGB7 |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | CSAD RARG SOAT2 ZNF740 ITGB7 |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | CSAD IGFBP6 RARG TNS2 SOAT2 SPRYD3 ZNF740 ITGB7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |