Assay Details
Target Gene Details
Entrez Gene ID: | 80185 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
C8orf41, MRT39 |
Location: |
Chr.8:33498509-33513601 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTI2 | NM_001102401.2 | NP_001095871.1 | ||
NM_001265581.1 | NP_001252510.1 | |||
NM_025115.3 | NP_079391.2 | |||
XM_011544665.2 | XP_011542967.1 | |||
AK026916.1 | BAB15590.1 | |||
BC007387.2 | AAH07387.1 | |||
BC066935.1 | AAH66935.1 | |||
BX648410.1 |