Assay Details
Target Gene Details
Entrez Gene ID: | 5099 |
Gene Name: | protocadherin 7 |
Gene Aliases: |
BH-Pcdh, BHPCDH, PPP1R120 |
Location: |
Chr.4:30720378-31146801 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCDH7 | NM_001173523.1 | 1 | 207 | NP_001166994.1 |
NM_002589.2 | 1 | 207 | NP_002580.2 | |
NM_032456.2 | 1 | 214 | NP_115832.1 | |
NM_032457.3 | 1 | 207 | NP_115833.2 | |
XM_005248163.3 | 1 | 242 | XP_005248220.1 | |
XM_005248164.3 | 1 | 242 | XP_005248221.1 | |
XM_005248166.3 | 1 | 241 | XP_005248223.1 | |
XM_011513842.2 | 1 | 243 | XP_011512144.1 | |
XM_011513843.2 | 1 | 242 | XP_011512145.1 | |
XM_011513844.2 | 1 | 243 | XP_011512146.1 | |
XM_017008259.1 | 1 | 243 | XP_016863748.1 | |
XM_017008260.1 | 1 | 243 | XP_016863749.1 | |
XM_017008261.1 | 1 | 243 | XP_016863750.1 | |
XM_017008262.1 | 1 | 243 | XP_016863751.1 | |
XM_017008263.1 | 1 | 244 | XP_016863752.1 | |
XM_017008264.1 | 1 | 244 | XP_016863753.1 | |
XM_017008265.1 | 1 | 244 | XP_016863754.1 | |
XM_017008266.1 | 1 | 244 | XP_016863755.1 | |
XM_017008267.1 | 1 | 244 | XP_016863756.1 | |
XM_017008268.1 | 1 | 244 | XP_016863757.1 | |
XM_017008269.1 | 1 | 244 | XP_016863758.1 | |
XM_017008270.1 | 1 | 243 | XP_016863759.1 | |
XM_017008271.1 | 1 | 243 | XP_016863760.1 | |
XM_017008272.1 | 1 | 242 | XP_016863761.1 | |
AB006755.1 | 1 | 207 | BAA25194.1 | |
AB006757.1 | BAA25196.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |