Assay Details
Target Gene Details
Entrez Gene ID: | 56474 |
Gene Name: | CTP synthase 2 |
Gene Aliases: |
- |
Location: |
Chr.X:16587999-16712979 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTPS2 | NM_001144002.1 | NP_001137474.1 | ||
NM_019857.4 | NP_062831.3 | |||
NM_175859.2 | NP_787055.1 | |||
XM_005274562.3 | XP_005274619.1 | |||
XM_006724503.3 | XP_006724566.1 | |||
XM_011545545.2 | XP_011543847.1 | |||
AF226667.1 | AAF91241.1 | |||
AK023549.1 | BAB14607.1 | |||
AK024070.1 | BAB14814.1 | |||
AK025654.1 | BAB15204.1 | |||
AK125332.1 | ||||
AK125348.1 | ||||
BC006256.2 | AAH06256.2 | |||
BC034986.1 | AAH34986.1 |
Target Gene Details
Entrez Gene ID: | 795 |
Gene Name: | S100 calcium binding protein G |
Gene Aliases: |
CABP, CABP1, CABP9K, CALB3 |
Location: |
Chr.X:16649787-16654674 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100G | NM_004057.2 | NP_004048.1 | ||
XM_017029841.1 | XP_016885330.1 | |||
BC112174.1 | AAI12175.1 | |||
L13220.1 | AAA35638.1 | |||
X65869.1 | CAA46699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6816 | Chr.X:16650590 - 16681559 on Build GRCh38 | Insertion | CTPS2 S100G |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |