Assay Details
Target Gene Details
Entrez Gene ID: | 8763 |
Gene Name: | CD164 molecule |
Gene Aliases: |
DFNA66, MGC-24, MUC-24, endolyn |
Location: |
Chr.6:109366514-109382559 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD164 | NM_001142401.1 | 5 | 1432 | NP_001135873.1 |
NM_001142402.1 | 5 | 1414 | NP_001135874.1 | |
NM_001142403.1 | 7 | 875 | NP_001135875.1 | |
NM_001142404.1 | 6 | 779 | NP_001135876.1 | |
NM_006016.4 | 6 | 1471 | NP_006007.2 | |
AF299341.1 | 6 | 1290 | AAG53906.1 | |
AF299342.1 | 5 | 1311 | AAG53907.1 | |
AF299343.1 | 5 | 1329 | AAG53908.1 | |
AK298684.1 | 6 | 631 | ||
AK301692.1 | ||||
AK303525.1 | 5 | 1266 | ||
AK307193.1 | ||||
AK310854.1 | 6 | 1312 | ||
AK315908.1 | 5 | 1283 | ||
BC011522.2 | 6 | 1371 | AAH11522.1 | |
D14043.1 | 7 | 773 | BAA03130.1 | |
EB388947.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |