Assay Details
Target Gene Details
Entrez Gene ID: | 6285 |
Gene Name: | S100 calcium binding protein B |
Gene Aliases: |
NEF, S100, S100-B, S100beta |
Location: |
Chr.21:46598618-46605242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100B | NM_006272.2 | NP_006263.1 | ||
XM_017028424.1 | XP_016883913.1 | |||
AA310307.1 | ||||
BC001766.1 | AAH01766.1 | |||
EB386131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528495 | Chr.21:46595956 - 46632926 on Build GRCh38 | Loss | S100B |
dgv4442n100 | Chr.21:46551020 - 46677033 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
nsv587999 | Chr.21:46522003 - 46683939 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
nsv588002 | Chr.21:46575530 - 46653004 on Build GRCh38 | Gain | S100B PRMT2 |
esv34393 | Chr.21:46595759 - 46657900 on Build GRCh38 | Gain | S100B PRMT2 |
esv3647171 | Chr.21:46586178 - 46674643 on Build GRCh38 | Gain | S100B PRMT2 |
dgv4443n100 | Chr.21:46561965 - 46646565 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
esv2758822 | Chr.21:46566757 - 46657900 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
dgv4441n100 | Chr.21:46474792 - 46677033 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
nsv528947 | Chr.21:46566170 - 46630476 on Build GRCh38 | Gain | S100B DIP2A |
esv3647172 | Chr.21:46592879 - 46650385 on Build GRCh38 | Loss | S100B PRMT2 |
dgv4444n100 | Chr.21:46599898 - 46677033 on Build GRCh38 | Gain | S100B PRMT2 |
esv2760700 | Chr.21:46554142 - 46634864 on Build GRCh38 | Gain | S100B DIP2A |
nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain | S100B LSS MCM3AP-AS1 FTCD PCNT SPATC1L LOC101928796 DIP2A-IT1 PRMT2 MCM3AP C21orf58 PCBP3 DIP2A YBEY COL6A2 COL6A1 |
nsv1065227 | Chr.21:46565742 - 46616312 on Build GRCh38 | Gain | S100B DIP2A |
dgv4446n100 | Chr.21:46601742 - 46654452 on Build GRCh38 | Gain | S100B PRMT2 |
nsv1057566 | Chr.21:46495670 - 46609786 on Build GRCh38 | Loss | S100B DIP2A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |