Assay Details
Target Gene Details
Entrez Gene ID: | 10094 |
Gene Name: | actin related protein 2/3 complex subunit 3 |
Gene Aliases: |
ARC21, p21-Arc |
Location: |
Chr.12:110434890-110450411 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARPC3 | NM_001278556.1 | NP_001265485.1 | ||
NM_001287222.1 | NP_001274151.1 | |||
AF004561.1 | AAB61466.1 | |||
AF006086.1 | AAB64191.1 | |||
AI144468.1 | ||||
AK297309.1 | ||||
AK311789.1 | ||||
AL357196.1 | ||||
BC067747.1 | AAH67747.1 | |||
BC078162.1 | AAH78162.1 | |||
BG620669.1 | ||||
BG723870.1 | ||||
BM999380.1 | ||||
CR407667.1 | CAG28595.1 | |||
DQ328220.1 | ABC59627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv976651 | Chr.12:110426752 - 110436265 on Build GRCh38 | Duplication | ARPC3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |