Assay Details
Target Gene Details
Entrez Gene ID: | 50861 |
Gene Name: | stathmin 3 |
Gene Aliases: |
SCLIP |
Location: |
Chr.20:63639705-63653610 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STMN3 | NM_015894.3 | NP_056978.2 | ||
NR_075070.1 | ||||
AK074860.1 | BAC11252.1 | |||
AK094112.1 | ||||
AK310010.1 | ||||
BC009381.2 | AAH09381.1 | |||
BC025234.1 | AAH25234.1 | |||
DA161621.1 | ||||
DA181040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953308 | Chr.20:63650148 - 63831847 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160690 | Chr.20:63639552 - 63744287 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv9827 | Chr.20:63648841 - 63653792 on Build GRCh38 | Loss |
![]() |
nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470563 | Chr.20:63486715 - 63793308 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758806 | Chr.20:63470996 - 63745014 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)