Assay Details
Target Gene Details
Entrez Gene ID: | 2831 |
Gene Name: | neuropeptides B/W receptor 1 |
Gene Aliases: |
GPR7 |
Location: |
Chr.8:52939908-52940894 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPBWR1 | NM_005285.3 | 1 | 284 | NP_005276.2 |
BC069117.1 | 1 | 284 | AAH69117.1 | |
BC107101.2 | 1 | 284 | AAI07102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759615 | Chr.8:52748233 - 53088532 on Build GRCh38 | Gain+Loss | NPBWR1 |
nsv1016224 | Chr.8:52807381 - 53454974 on Build GRCh38 | Gain | NPBWR1 LOC100507516 OPRK1 |
esv2764065 | Chr.8:52452089 - 52975372 on Build GRCh38 | Gain | FAM150A NPBWR1 LOC107986877 RB1CC1 |
esv3891397 | Chr.8:52798886 - 53126161 on Build GRCh38 | Gain | NPBWR1 |
nsv1023316 | Chr.8:52853703 - 52980958 on Build GRCh38 | Gain | NPBWR1 |
nsv824633 | Chr.8:52827411 - 52953769 on Build GRCh38 | Gain | NPBWR1 |
nsv525238 | Chr.8:52921934 - 53029113 on Build GRCh38 | Gain | NPBWR1 |
esv3617224 | Chr.8:52910629 - 52967647 on Build GRCh38 | Gain | NPBWR1 |
nsv1032190 | Chr.8:52893043 - 52984766 on Build GRCh38 | Gain | NPBWR1 |
dgv7218n100 | Chr.8:52389530 - 53007050 on Build GRCh38 | Gain | FAM150A NPBWR1 LOC107986877 RB1CC1 ST18 |
nsv1016750 | Chr.8:52932614 - 53020512 on Build GRCh38 | Gain | NPBWR1 |
dgv7222n100 | Chr.8:52753073 - 52968653 on Build GRCh38 | Gain | NPBWR1 |
esv3576572 | Chr.8:52429401 - 52997384 on Build GRCh38 | Gain | FAM150A NPBWR1 LOC107986877 RB1CC1 |
nsv8344 | Chr.8:52938312 - 52941949 on Build GRCh38 | Loss | NPBWR1 |
esv3617225 | Chr.8:52938447 - 53031293 on Build GRCh38 | Gain | NPBWR1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |