Assay Details
Target Gene Details
Entrez Gene ID: | 253152 |
Gene Name: | epoxide hydrolase 4 |
Gene Aliases: |
ABHD7, EH4, EPHXRP |
Location: |
Chr.1:92029976-92063536 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHX4 | NM_173567.4 | NP_775838.3 | ||
AK074822.1 | BAC11230.1 | |||
BC041475.1 | AAH41475.1 | |||
DB475492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002730 | Chr.1:91836984 - 92099672 on Build GRCh38 | Gain | BRDT EPHX4 TGFBR3 SETSIP BTBD8 |
nsv524845 | Chr.1:91750168 - 92133148 on Build GRCh38 | Gain | BRDT EPHX4 TGFBR3 SETSIP BTBD8 |
nsv830581 | Chr.1:91851650 - 92045618 on Build GRCh38 | Loss | BRDT EPHX4 TGFBR3 |
esv2750842 | Chr.1:91891224 - 92114118 on Build GRCh38 | Gain | BRDT EPHX4 TGFBR3 SETSIP BTBD8 |
esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion | C1orf146 HFM1 SNORA66 TGFBR3 EVI5 SNORD21 SETSIP BTBD8 FAM69A BRDT EPHX4 KIAA1107 GFI1 RPL5 RPAP2 HSP90B3P GLMN CDC7 |
nsv1006318 | Chr.1:91750851 - 92142767 on Build GRCh38 | Gain | BRDT EPHX4 TGFBR3 SETSIP BTBD8 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |