Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | 4 | 1855 | NP_005348.2 |
XM_005258937.3 | 4 | 1850 | XP_005258994.1 | |
XM_005258938.4 | 4 | 841 | XP_005258995.1 | |
XM_005258939.3 | 5 | 879 | XP_005258996.2 | |
XM_005258940.3 | 4 | 780 | XP_005258997.1 | |
XM_006723218.2 | 4 | 980 | XP_006723281.1 | |
XM_017026810.1 | 4 | 720 | XP_016882299.1 | |
AK292482.1 | 4 | 1835 | ||
BC070041.1 | 4 | 1855 | AAH70041.1 | |
U40002.1 | 4 | 1855 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 | ||||
DW455857.1 |
Target Gene Details
Entrez Gene ID: | 101930071 |
Gene Name: | uncharacterized LOC101930071 |
Gene Aliases: |
- |
Location: |
Chr.19:42397128-42408452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101930071 | NR_126041.1 | |||
BU566442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | LOC101930071 LIPE LIPE-AS1 CNFN |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | CD79A MIR6797 ATP1A3 POU2F2 MIR8077 LOC101930071 GRIK5 LOC100505622 LOC100505585 ARHGEF1 MEGF8 DEDD2 ZNF526 ZNF574 PRR19 PAFAH1B3 LOC390937 CIC RPS19 TMEM145 MIR4323 RABAC1 GSK3A LIPE LIPE-AS1 CNFN ERF |
nsv579632 | Chr.19:42400501 - 42408251 on Build GRCh38 | Gain | LOC101930071 LIPE LIPE-AS1 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | PAFAH1B3 POU2F2 MIR8077 LOC101930071 CXCL17 CEACAM8 CIC MEGF8 DEDD2 TMEM145 GSK3A ZNF526 LIPE LIPE-AS1 PRR19 CEACAM1 CNFN ERF |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | CD79A MIR6797 ATP1A3 POU2F2 MIR8077 CEACAM4 LOC101930071 GRIK5 LOC100505622 LOC100505585 ARHGEF1 MEGF8 DEDD2 ZNF526 ZNF574 PRR19 PAFAH1B3 CEACAM5 CEACAM3 CEACAM7 LOC390937 LYPD4 CIC RPS19 TMEM145 MIR4323 RABAC1 GSK3A DMRTC2 CEACAM6 LIPE LIPE-AS1 CNFN ERF |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | LOC100289650 PAFAH1B3 PSG10P PSG1 MIR8077 PSG11 LOC101930071 CXCL17 PSG8 PSG2 CEACAM8 CIC MEGF8 TMEM145 PSG3 GSK3A PSG7 PSG6 LIPE LIPE-AS1 PRR19 CEACAM1 CNFN ERF |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |