Assay Details
Target Gene Details
Entrez Gene ID: | 7075 |
Gene Name: | tyrosine kinase with immunoglobulin like and EGF like domains 1 |
Gene Aliases: |
JTK14, TIE |
Location: |
Chr.1:43300895-43323110 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TIE1 | NM_001253357.1 | 13 | 2087 | NP_001240286.1 |
NM_005424.4 | 13 | 2170 | NP_005415.1 | |
XM_005271163.1 | 12 | 1984 | XP_005271220.1 | |
XM_006710869.1 | 13 | 2304 | XP_006710932.1 | |
XM_017002207.1 | 13 | 2113 | XP_016857696.1 | |
AF217976.1 | 3 | 254 | AAG17219.1 | |
AK225560.1 | 13 | 1829 | ||
AK296735.1 | 8 | 1146 | ||
AK300399.1 | 13 | 1977 | ||
AK300623.1 | 13 | 1870 | ||
AK307639.1 | 6 | 838 | ||
BC038239.1 | 13 | 2072 | AAH38239.1 | |
EU826590.1 | 13 | 1993 | ||
X60957.1 | 13 | 2029 | CAA43290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss | CDC20 MIR6735 SZT2 HYI MPL ELOVL1 TIE1 MED8 MIR6734 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |