Assay Details
Target Gene Details
Entrez Gene ID: | 144100 |
Gene Name: | pleckstrin homology domain containing A7 |
Gene Aliases: |
- |
Location: |
Chr.11:16787153-17014423 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLEKHA7 | NM_175058.4 | 23 | 3233 | NP_778228.3 |
XM_017017240.1 | 25 | 3444 | XP_016872729.1 | |
XM_017017241.1 | 25 | 3444 | XP_016872730.1 | |
XM_017017242.1 | 24 | 3324 | XP_016872731.1 | |
XM_017017243.1 | 23 | 3243 | XP_016872732.1 | |
AK296371.1 | 13 | 1971 | ||
AK301465.1 | 13 | 1827 | ||
BC033239.1 | 14 | 2113 | AAH33239.1 | |
BC041452.1 | 5 | 571 | AAH41452.1 | |
BC071599.1 | 23 | 3229 | AAH71599.1 | |
BX648885.1 | 2 | 2016 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |