Assay Details
Target Gene Details
Entrez Gene ID: | 51726 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B11 |
Gene Aliases: |
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537 |
Location: |
Chr.3:186570676-186585800 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNAJB11 | NM_016306.5 | 2 | 399 | NP_057390.1 |
AB028859.2 | 2 | 342 | BAA88307.1 | |
AF228505.1 | 2 | 268 | AAF61711.1 | |
AF277317.1 | 2 | 359 | AAK69110.1 | |
AJ250137.1 | 2 | 197 | CAB65118.1 | |
AK075300.1 | 2 | 356 | BAC11533.1 | |
AK075430.1 | 2 | 375 | BAC11617.1 | |
AK301292.1 | ||||
AY359043.1 | 2 | 335 | AAQ89402.1 | |
BC001144.1 | 2 | 337 | AAH01144.1 | |
BM754506.1 | 2 | 386 | ||
BT007063.1 | 2 | 177 | AAP35712.1 | |
CR457096.1 | 2 | 177 | CAG33377.1 | |
DA058195.1 | 2 | 399 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |