Assay Details
Target Gene Details
Entrez Gene ID: | 84310 |
Gene Name: | chromosome 7 open reading frame 50 |
Gene Aliases: |
YCR016W |
Location: |
Chr.7:978485-1138291 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C7orf50 | XM_011515580.2 | XP_011513882.1 | ||
XM_011515581.2 | XP_011513883.1 | |||
XM_011515582.2 | XP_011513884.1 | |||
XM_011515583.2 | XP_011513885.1 | |||
XM_011515584.2 | XP_011513886.1 | |||
XM_017012719.1 | XP_016868208.1 | |||
XM_017012720.1 | XP_016868209.1 | |||
XM_017012721.1 | XP_016868210.1 |
Target Gene Details
Entrez Gene ID: | 54905 |
Gene Name: | cytochrome P450 family 2 subfamily W member 1 |
Gene Aliases: |
- |
Location: |
Chr.7:983157-989640 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP2W1 | NM_017781.2 | NP_060251.2 | ||
XM_011515440.2 | XP_011513742.1 | |||
XM_011515441.2 | XP_011513743.1 | |||
AK000366.1 | BAA91114.1 | |||
BC025761.1 | AAH25761.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv605720 | Chr.7:977312 - 1121522 on Build GRCh38 | Loss | LOC107986755 C7orf50 GPER1 GPR146 CYP2W1 MIR339 |
nsv605700 | Chr.7:886440 - 1065790 on Build GRCh38 | Gain | LOC107986755 GET4 C7orf50 COX19 ADAP1 GPR146 CYP2W1 MIR339 |
nsv482099 | Chr.7:983199 - 989640 on Build GRCh38 | Gain | C7orf50 CYP2W1 |
esv3571559 | Chr.7:981839 - 1000115 on Build GRCh38 | Loss | C7orf50 CYP2W1 |
nsv605699 | Chr.7:886312 - 1073232 on Build GRCh38 | Loss | LOC107986755 GET4 C7orf50 COX19 ADAP1 GPR146 CYP2W1 MIR339 |
nsv1028282 | Chr.7:964887 - 1000659 on Build GRCh38 | Gain | C7orf50 COX19 CYP2W1 |
nsv509182 | Chr.7:881218 - 1018010 on Build GRCh38 | Insertion | GET4 C7orf50 COX19 ADAP1 CYP2W1 |
nsv950784 | Chr.7:855964 - 1074664 on Build GRCh38 | Deletion | LOC107986755 GET4 C7orf50 COX19 ADAP1 GPR146 CYP2W1 MIR339 SUN1 |
esv3571557 | Chr.7:823644 - 994725 on Build GRCh38 | Loss | GET4 C7orf50 COX19 ADAP1 CYP2W1 LOC107986754 SUN1 |
nsv605705 | Chr.7:947983 - 1211575 on Build GRCh38 | Loss | LOC101927021 LOC107986755 C7orf50 ZFAND2A COX19 ADAP1 GPER1 GPR146 CYP2W1 MIR339 |
nsv1161477 | Chr.7:928825 - 1077800 on Build GRCh38 | Deletion | LOC107986755 C7orf50 COX19 ADAP1 GPR146 CYP2W1 MIR339 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |