Assay Details
Target Gene Details
Entrez Gene ID: | 1048 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 5 |
Gene Aliases: |
CD66e, CEA |
Location: |
Chr.19:41708585-41730433 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM5 | NM_001291484.2 | NP_001278413.1 | ||
NM_001308398.1 | NP_001295327.1 | |||
NM_004363.5 | NP_004354.3 | |||
XM_005258413.3 | XP_005258470.2 | |||
XM_011526322.1 | XP_011524624.1 | |||
XM_017026145.1 | XP_016881634.1 | |||
XM_017026146.1 | XP_016881635.1 | |||
AB852566.1 | ||||
AB852567.1 | ||||
AK223101.1 | BAD96821.1 | |||
BC034671.1 | AAH34671.1 | |||
CR749337.1 | CAH18191.1 | |||
M15042.1 | AAA51963.1 | |||
M29540.1 | AAA51967.1 | |||
X16455.1 | CAA34474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv963052 | Chr.19:41714363 - 41721068 on Build GRCh38 | Duplication | CEACAM5 |
nsv1061309 | Chr.19:41683785 - 41724525 on Build GRCh38 | Gain | CEACAM5 CEACAM7 |
nsv482220 | Chr.19:41708611 - 41730516 on Build GRCh38 | Gain | CEACAM5 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | DMRTC2 ARHGEF1 GRIK5 CIC MEGF8 GSK3A POU2F2 LOC101930071 CEACAM5 CEACAM6 RPS19 LOC100505585 TMEM145 MIR4323 LIPE ERF CEACAM3 LYPD4 CNFN PRR19 CEACAM4 MIR6797 RABAC1 ZNF574 LOC100505622 LOC390937 PAFAH1B3 ZNF526 ATP1A3 DEDD2 LIPE-AS1 CEACAM7 MIR8077 CD79A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |