Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | NP_057356.1 | ||
XM_005261315.2 | XP_005261372.1 | |||
XM_006724105.3 | XP_006724168.1 | |||
XM_017028539.1 | XP_016884028.1 | |||
AB035207.1 | 1 | 194 | BAA87042.1 | |
AB051450.1 | ||||
BC028919.1 | AAH28919.1 | |||
BC038957.1 | AAH38957.1 | |||
BM453650.1 | ||||
CR456594.1 | CAG30480.1 | |||
CR541684.1 | CAG46485.1 | |||
D64109.1 | 1 | 73 | BAA10971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss | TOB2 TEF ZC3H7B PHF5A RANGAP1 LOC100996598 |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss | ACO2 TOB2 TEF CSDC2 POLR3H ZC3H7B LOC105373044 PHF5A MIR6889 PMM1 RANGAP1 LOC100996598 |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss | ACO2 TOB2 TEF POLR3H LOC105373044 PHF5A LOC100996598 |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain | TOB2 TEF ZC3H7B MIR6889 CHADL RANGAP1 LOC100996598 |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | SNORD140 EP300 L3MBTL2 POLR3H LOC105373044 PHF5A MIR6889 CHADL PMM1 RANGAP1 LOC100996598 DESI1 SNU13 ACO2 EP300-AS1 TOB2 TEF MEI1 CSDC2 ZC3H7B XRCC6 C22orf46 MIR1281 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |