Assay Details
Target Gene Details
Entrez Gene ID: | 143 |
Gene Name: | poly(ADP-ribose) polymerase family member 4 |
Gene Aliases: |
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193 |
Location: |
Chr.13:24420930-24512993 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARP4 | NM_006437.3 | 31 | 4027 | NP_006428.2 |
XM_011534931.1 | 30 | 4077 | XP_011533233.1 | |
XM_011534932.2 | 31 | 4244 | XP_011533234.1 | |
AF057160.1 | 31 | 3973 | AAC62491.1 | |
AF158255.1 | 31 | 4027 | AAD47250.1 | |
D79999.2 | 30 | 3926 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832567 | Chr.13:24302981 - 24520733 on Build GRCh38 | Loss | LINC00566 SPATA13 PARP4 C1QTNF9 NUS1P3 |
esv3892310 | Chr.13:24212887 - 24577193 on Build GRCh38 | Gain | LINC00566 LOC105370295 SPATA13 SPATA13-AS1 PARP4 C1QTNF9 NUS1P3 LOC101927375 |
nsv1036613 | Chr.13:24347567 - 24461469 on Build GRCh38 | Gain | PARP4 |
nsv977216 | Chr.13:24398558 - 24444213 on Build GRCh38 | Duplication | PARP4 |
dgv1616n100 | Chr.13:24367378 - 24455157 on Build GRCh38 | Gain | PARP4 |
esv3582215 | Chr.13:24193682 - 24455263 on Build GRCh38 | Gain | LINC00566 SPATA13 SPATA13-AS1 PARP4 C1QTNF9 NUS1P3 |
esv3631556 | Chr.13:24426828 - 24487208 on Build GRCh38 | Loss | PARP4 |
esv2761200 | Chr.13:24313097 - 24636778 on Build GRCh38 | Gain+Loss | LINC00566 LOC105370295 PARP4 C1QTNF9 NUS1P3 TPTE2P6 LOC101927375 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |