Assay Details
Target Gene Details
Entrez Gene ID: | 4717 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit C1 |
Gene Aliases: |
KFYI |
Location: |
Chr.4:139289917-139302551 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFC1 | NM_001184986.1 | 6 | 1139 | NP_001171915.1 |
NM_001184987.1 | 5 | 607 | NP_001171916.1 | |
NM_001184988.1 | 6 | 643 | NP_001171917.1 | |
NM_001184989.1 | 6 | 666 | NP_001171918.1 | |
NM_001184990.1 | 5 | 603 | NP_001171919.1 | |
NM_001184991.1 | 5 | 1080 | NP_001171920.1 | |
NM_002494.3 | 5 | 742 | NP_002485.1 | |
AF047184.1 | 4 | 337 | AAC04269.1 | |
AF047435.1 | 4 | 317 | AAC39886.1 | |
AK291147.1 | 4 | 347 | ||
BC101798.1 | AAI01799.1 | |||
BC107682.1 | 5 | 712 | AAI07683.1 | |
BC112061.1 | AAI12062.1 | |||
BG701467.1 | 5 | 747 | ||
BM768825.1 | ||||
BU677336.1 | 1 | 81 | ||
CA313899.1 | 1 | 118 | ||
CB047240.1 | 1 | 128 | ||
CB160001.1 | ||||
CN347371.1 | 5 | 508 | ||
W56292.1 | 1 | 119 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |