Assay Details
Target Gene Details
Entrez Gene ID: | 255189 |
Gene Name: | phospholipase A2 group IVF |
Gene Aliases: |
PLA2G4FZ |
Location: |
Chr.15:42141134-42156641 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G4F | NM_213600.3 | 20 | 3309 | NP_998765.3 |
NR_033151.1 | 19 | 3236 | ||
AK122623.1 | 21 | 2920 | ||
AK172836.1 | BAD18801.1 | |||
AK304368.1 | 20 | 3291 | ||
AL832972.1 | 3 | 1283 | ||
BC035335.1 | 19 | 3206 | AAH35335.1 | |
BC131531.1 | 20 | 3221 | ||
BC142682.1 | 20 | 3280 | ||
BC146648.1 | 20 | 3280 | ||
BM986790.1 | 1 | 135 | ||
CR749451.1 | 13 | 5382 | CAH18288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3636260 | Chr.15:42129273 - 42141661 on Build GRCh38 | Loss | PLA2G4F |
nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain | PLA2G4B PLA2G4D JMJD7 MAPKBP1 PLA2G4E-AS1 MIR4310 EHD4-AS1 PLA2G4E JMJD7-PLA2G4B PLA2G4F SPTBN5 EHD4 LOC105370792 |
esv2749622 | Chr.15:42129196 - 42141609 on Build GRCh38 | Deletion | PLA2G4F |
esv2664756 | Chr.15:42129205 - 42141617 on Build GRCh38 | Deletion | PLA2G4F |
esv3385504 | Chr.15:42140662 - 42142560 on Build GRCh38 | Duplication | PLA2G4F |
nsv952614 | Chr.15:42139503 - 42164202 on Build GRCh38 | Deletion | PLA2G4F VPS39 |
nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss | PLA2G4E PLA2G4D TMEM87A PLA2G4F MIR627 VPS39 |
nsv442371 | Chr.15:42129217 - 42141608 on Build GRCh38 | Loss | PLA2G4F |
nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss | PLA2G4D TMEM87A PLA2G4F MIR627 VPS39 |
esv3892678 | Chr.15:42130892 - 42142055 on Build GRCh38 | Loss | PLA2G4F |
nsv569237 | Chr.15:42125887 - 42145233 on Build GRCh38 | Loss | PLA2G4F |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |