Assay Details
Target Gene Details
Entrez Gene ID: | 4750 |
Gene Name: | NIMA related kinase 1 |
Gene Aliases: |
NY-REN-55, SRPS2, SRPS2A, SRTD6 |
Location: |
Chr.4:169393270-169612629 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEK1 | NM_001199397.1 | 36 | 4986 | NP_001186326.1 |
NM_001199398.1 | 35 | 4854 | NP_001186327.1 | |
NM_001199399.1 | 33 | 4695 | NP_001186328.1 | |
NM_001199400.1 | 33 | 4848 | NP_001186329.1 | |
NM_012224.2 | 34 | 4980 | NP_036356.1 | |
XM_006714228.1 | 35 | 4811 | XP_006714291.1 | |
XM_011532003.1 | 35 | 4904 | XP_011530305.1 | |
XM_011532004.1 | 34 | 4772 | XP_011530306.1 | |
XM_017008249.1 | 35 | 4890 | XP_016863738.1 | |
XM_017008250.1 | 34 | 5008 | XP_016863739.1 | |
XM_017008251.1 | 34 | 4806 | XP_016863740.1 | |
XM_017008252.1 | 32 | 4826 | XP_016863741.1 | |
XM_017008253.1 | 35 | 4836 | XP_016863742.1 | |
XM_017008254.1 | 28 | 3946 | XP_016863743.1 | |
AB067488.1 | 34 | 4816 | ||
AF155113.1 | 18 | 2864 | AAD42879.1 | |
AK025658.1 | 9 | 1734 | BAB15207.1 | |
AL050385.1 | 35 | 4810 | CAI46225.1 | |
BC037790.2 | 8 | 1646 | AAH37790.1 | |
CR933642.1 | 36 | 4944 | CAI45943.1 | |
CR936672.1 | 33 | 4806 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |