Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOCS2 | NM_001270467.1 | 3 | 1189 | NP_001257396.1 |
NM_001270468.1 | 3 | 1158 | NP_001257397.1 | |
NM_001270469.1 | 3 | 1156 | NP_001257398.1 | |
NM_001270470.1 | 3 | 1428 | NP_001257399.1 | |
NM_001270471.1 | 2 | 1081 | NP_001257400.1 | |
NM_003877.4 | 3 | 1299 | NP_003868.1 | |
XM_011538929.1 | XP_011537231.1 | |||
XM_011538935.1 | XP_011537237.1 | |||
XM_011538936.1 | XP_011537238.1 | |||
XM_017020147.1 | XP_016875636.1 | |||
XM_017020148.1 | XP_016875637.1 | |||
XM_017020149.1 | XP_016875638.1 | |||
XM_017020150.1 | XP_016875639.1 | |||
XM_017020151.1 | XP_016875640.1 | |||
XM_017020152.1 | XP_016875641.1 | |||
XM_017020153.1 | XP_016875642.1 | |||
XM_017020154.1 | XP_016875643.1 | |||
XM_017020155.1 | XP_016875644.1 | |||
XM_017020156.1 | 3 | 1414 | XP_016875645.1 | |
AB004903.1 | BAA22429.1 | |||
AF020590.1 | 2 | 908 | AAC98896.1 | |
AF037989.1 | 3 | 1026 | AAC34745.1 | |
AK290546.1 | ||||
AK297662.1 | 4 | 889 | ||
AK309452.1 | 3 | 695 | ||
BC010399.1 | 2 | 921 | AAH10399.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |