Assay Details
Target Gene Details
Entrez Gene ID: | 11113 |
Gene Name: | citron rho-interacting serine/threonine kinase |
Gene Aliases: |
CRIK, STK21 |
Location: |
Chr.12:119685790-119877318 on Build GRCh38 |
Assay Gene Location: | Within Exon 45 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIT | NM_001206999.1 | 43 | 5554 | NP_001193928.1 |
NM_007174.2 | 42 | 5428 | NP_009105.1 | |
XM_006719206.2 | 43 | 5536 | XP_006719269.1 | |
XM_011537783.1 | 44 | 5626 | XP_011536085.1 | |
XM_011537784.1 | 44 | 5626 | XP_011536086.1 | |
XM_011537785.1 | 44 | 5581 | XP_011536087.1 | |
XM_011537787.1 | 42 | 5410 | XP_011536089.1 | |
XM_011537788.1 | 42 | 5410 | XP_011536090.1 | |
XM_011537789.1 | 36 | 4761 | XP_011536091.1 | |
XM_011537790.2 | 35 | 4329 | XP_011536092.1 | |
XM_011537791.2 | 35 | 5314 | XP_011536093.1 | |
XM_011537792.2 | 34 | 5369 | XP_011536094.1 | |
XM_017018735.1 | 43 | 5581 | XP_016874224.1 | |
XM_017018736.1 | 44 | 5581 | XP_016874225.1 | |
XM_017018737.1 | 43 | 5536 | XP_016874226.1 | |
XM_017018738.1 | 33 | 4870 | XP_016874227.1 | |
XM_017018739.1 | 33 | 5645 | XP_016874228.1 | |
AB023166.2 | 34 | 5369 | BAA76793.2 | |
AY209000.1 | AAP43922.1 | |||
AY257469.1 | 42 | 5425 | AAP13528.1 | |
AY681966.1 | 43 | 5554 | AAV87216.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |