Assay Details
Target Gene Details
Entrez Gene ID: | 11096 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 5 |
Gene Aliases: |
ADAM-TS 11, ADAM-TS 5, ADAM-TS5, ADAMTS-11, ADAMTS-5, ADAMTS11, ADMP-2 |
Location: |
Chr.21:26917912-26967120 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAMTS5 | NM_007038.4 | 8 | 8978 | NP_008969.2 |
XM_011529448.2 | 7 | 8810 | XP_011527750.1 | |
AK095905.1 | 1 | 1955 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1067026 | Chr.21:26910341 - 26925730 on Build GRCh38 | Gain | ADAMTS5 |
nsv834068 | Chr.21:26807351 - 26988489 on Build GRCh38 | Loss | MIR4759 ADAMTS5 ADAMTS1 LOC107987291 |
nsv1066573 | Chr.21:26724037 - 27002035 on Build GRCh38 | Gain | MIR4759 ADAMTS5 ADAMTS1 LOC107987291 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |