Assay Details
Target Gene Details
Entrez Gene ID: | 10463 |
Gene Name: | solute carrier family 30 member 9 |
Gene Aliases: |
C4orf1, GAC63, HUEL, ZNT9 |
Location: |
Chr.4:41990472-42087534 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC30A9 | NM_006345.3 | 5 | 628 | NP_006336.3 |
XM_017007654.1 | 5 | 662 | XP_016863143.1 | |
AF006621.2 | 5 | 602 | AAB87763.2 | |
AK225470.1 | 5 | 565 | ||
AK299917.1 | 6 | 721 | ||
AK312838.1 | 5 | 628 | ||
AY319413.1 | 6 | 507 | AAP83846.1 | |
AY594282.1 | 5 | 482 | AAT02479.1 | |
BC007732.2 | 5 | 589 | AAH07732.1 | |
BC016949.1 | 5 | 607 | AAH16949.1 | |
BC022981.1 | AAH22981.1 | |||
KU178526.1 | 5 | 482 | ||
KU178527.1 | 5 | 482 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |